(±)-2-Butanol ≥99%, ReagentPlus®

Danger

Synonyms: (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol

B85919-4L B85919-20L-P2 B85919-1L
220112-670EA 57.54 USD
220112-670 220112-669 220112-667
(±)-2-Butanol ≥99%, ReagentPlus®
(±)-2-Butanol
Formula: H₃CCH₂CH(OH)CH₃
MW: 74.12 g/mol
Boiling Pt: 94 °C (1013 hPa)
Melting Pt: -115 °C
Density: 0.8534 g/cm³ (20 °C)
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00004569
CAS Number: 78-92-2
EINECS: 201-158-5
UN: 1120
ADR: 3,III
Merck Index: 13,01540
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dea3607167631fbfcf7751c20b71f345ad24c19&vid=93e0b6b0d66ee031081607904a64140d92635df1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR