1,4-Butanesultone

Synonyms: 1,4-Butane sultone

B85501-100G B85501-25G
220112-659EA 149.69 USD
220112-659 220112-660
1,4-Butanesultone
1,4-Butanesultone
Formula: C₄H₈O₃S
MW: 136.17 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: 12.5…15 °C
Density: 1.331 g/cm³ (20 °C)
MDL Number: MFCD00006584
CAS Number: 1633-83-6
EINECS: 216-647-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR