(±)-1,3-Butanediol ≥99%, ReagentPlus®

Synonyms: Butane-1,3-diol, (±)-1,3-Butylene glycol, (±)-1,3-Dihydroxy butane

B84785-4L B84785-1L B84785-100ML
220111-531EA 236.15 USD
220111-531 220111-529 220111-528
(±)-1,3-Butanediol ≥99%, ReagentPlus®
(±)-1,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 206 - 208 °C (1013 hPa)
Melting Pt: ? -50 °C
Density: 1.004 to 1.006 g/cm³ (20 °C)
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00004554
CAS Number: 107-88-0
EINECS: 203-529-7
Merck Index: 13,01566

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR