Bromoacetic acid 97%, Reagent Grade

Danger

B56307-500G B56307-100G B56307-2KG
220111-903EA 70.62 USD
220111-903 220111-901 220111-902
Bromoacetic acid 97%, Reagent Grade
Bromoacetic acid
Formula: BrCH₂COOH
MW: 138.95 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: 46 - 49 °C
Density: 1.9335 g/cm³ (20 °C)
Flash Pt: ≥ 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00002678
CAS Number: 79-08-3
EINECS: 201-175-8
UN: 3425
ADR: 8,II
Merck Index: 13,01384

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR