Bis(tributyltin) oxide ≥96%

Danger

Synonyms: Bis(tri-n-butyltin)oxide, TBTO

B53383-100G B53383-500G
220111-870EA 259.02 USD
220111-870 220111-871
Bis(tributyltin) oxide ≥96%
Bis(tributyltin) oxide
Formula: C₂₄H₅₄OSn₂
MW: 596.11 g/mol
Boiling Pt: 475 °C (1013 hPa)
Density: 1.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009418
CAS Number: 56-35-9
EINECS: 200-268-0
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR