1,2-Diphenylethane ≥99%

Synonyms: Dibenzyl, Bibenzyl

B33706-25G B33706-100G
220111-489EA 108.77 USD
220111-489 220111-488
1,2-Diphenylethane ≥99%
1,2-Diphenylethane
Formula: (C₆H₅)CH₂CH₂(C₆H₅)
MW: 182.27 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 52 - 54 °C
Density: 1.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004796
CAS Number: 103-29-7
EINECS: 203-096-4
Merck Index: 12,01245

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR