Benzoyl chloride ≥99%, ReagentPlus®

Danger

Synonyms: Benzoic acid chloride

B12695-25ML B12695-1L-CB B12695-4L B12695-1L
220111-017EA 27.69 USD
220111-017 220111-016 220111-019 220111-015
Benzoyl chloride ≥99%, ReagentPlus®
Benzoyl chloride
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 197.2 °C (1013 hPa)
Melting Pt: ∼–0.6 °C
Density: 1.2023 g/cm³ (20 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00000653
CAS Number: 98-88-4
EINECS: 202-710-8
UN: 1736
ADR: 8,II
Merck Index: 13,01113

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR