p-Benzoquinone, Reagent Grade

Danger

Synonyms: p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone

B10358-5G B10358-1KG B10358-100G B10358-500G
220110-976EA 22.38 USD
220110-976 220110-974 220110-973 220110-975
p-Benzoquinone, Reagent Grade
p-Benzoquinone
Formula: C₆H₄O₂
MW: 108.1 g/mol
Boiling Pt: ∼174 °C (1013 hPa)
Melting Pt: 112…114 °C
Density: 1.318 g/cm³ (20 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00001591
CAS Number: 106-51-4
EINECS: 203-405-2
UN: 2587
ADR: 6.1,II
Merck Index: 12,08259
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=759b78f70866f47baad25776f322417b64521955&vid=331add73efa00881381cbace038e21cbca76318f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR