Butyric anhydride ≥98%

Danger

B103551-500ML B103551-1L
220110-500EA 72.69 USD
220110-500 220110-499
Butyric anhydride ≥98%
Butyric anhydride
Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: –73 °C
Density: 0.963 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00009389
CAS Number: 106-31-0
EINECS: 203-383-4
UN: 2739
ADR: 8,III
Merck Index: 13,01594

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR