7-Azaindole

Synonyms: 1H-Pyrrolo[2,3-b]pyridine

A95502-1G A95502-5G
220105-572EA 49.43 USD
220105-572 220105-573
7-Azaindole
7-Azaindole
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 103 - 106 °C
Density: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00005606
CAS Number: 271-63-6
EINECS: 205-981-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR