N-Acetyl-L-cysteine

Synonyms: Acetylcysteine, N-Acetyl-3-mercaptoalanine, NAC, (R)-(+)-N-Acetylcysteine, (R)-2-Acetamido-3-mercaptopropanoic acid, Ac-Cys-OH, N-Acetyl-L-Cysteine, LNAC

A7250-100G A7250-25G A7250-10G A7250-1KG A7250-5G A7250-50G A7250-500G
220104-434EA 267.08 USD
220104-434 220104-437 220104-435 220104-436 220104-440 220104-439 220104-438
N-Acetyl-L-cysteine
N-Acetyl-L-cysteine
Formula: C₅H₉NO₃S
MW: 163.2 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 106…108 °C
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004880
CAS Number: 616-91-1
EINECS: 210-498-3
Merck Index: 13,00090

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR