4-Amino-m-toluic acid ≥98%

Warning

Synonyms: 4-Amino-3-methylbenzoic acid

A63007-10G A63007-1G
220104-097EA 385.85 USD
220104-097 220104-098
4-Amino-m-toluic acid ≥98%
4-Amino-m-toluic acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 170 °C
MDL Number: MFCD00007736
CAS Number: 2486-70-6
EINECS: 219-629-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR