TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether) solution

X405-100ML X405-1L X405-1GA X405-500ML X405-5GA
220240-442EA 65.71 USD
220240-442 220240-444 220240-443 220240-445 220240-446
TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether) solution
TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether)
Formula: C₂₈H₅₀O₈
MW: 514.69 g/mol
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: –4 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779734
CAS Number: 9002-93-1
Merck Index: 12,06858

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR