Xylitol

X3375-10MG X3375-25G X3375-100G X3375-5G X3375-500G X3375-1KG
220239-605EA 17.23 USD
220239-605 220239-607 220239-604 220239-609 220239-608 220239-606
Xylitol
Xylitol
Formula: C₅H₁₂O₅
Boiling Pt: 495 °C (1013 hPa)
Melting Pt: 94 °C
Density: 1.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064292
CAS Number: 87-99-0
EINECS: 201-788-0
Merck Index: 13,10140

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR