L(+)-Isoleucine ≥98.5% FCC

Synonyms: L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH

W527602-1KG W527602-100G
220236-235EA 421.54 USD
220236-235 220236-234
L(+)-Isoleucine ≥98.5% FCC
L(+)-Isoleucine
Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17 g/mol
Melting Pt: 279 - 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS Number: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR