Coumarin ≥98%

Danger

W526509-10KG W526509-5KG W526509-1KG
220236-214EA 692.31 USD
220236-214 220236-216 220236-215
Coumarin ≥98%
Coumarin

Coumarin is an aroma compound responsible for the characteristic odor of hay. It is one of the major volatile compounds found in the aerial parts of bastard balm.


  • biological source: synthetic
  • grade: Halal

Formula: C₉H₆O₂
Boiling Pt: 298 °C (lit.)
Melting Pt: 67…73 °C (lit.)
Density: 0.935 g/cm³ (20 °C)
Flash Pt: 162 °C
Storage Temperature: Ambient
MDL Number: MFCD00006850
CAS Number: 91-64-5
EINECS: 202-086-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,02588

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR