Methyl-2,4-dihydroxy-3,6-dimethylbenzoate ≥98%

Synonyms: Methyl atratate, Atraric acid, Methyl atratat, Methyl 2,4-dihydroxy-3,6-dimethylbenzoate, 2,4-Dihydroxy-3,6-dimethylbenzoic acid methyl ester

W523305-1KG W523305-10KG
220235-825EA 452.31 USD
220235-825 220235-824
Methyl-2,4-dihydroxy-3,6-dimethylbenzoate ≥98%
Methyl-2,4-dihydroxy-3,6-dimethylbenzoate
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00157202
CAS Number: 4707-47-5
EINECS: 225-193-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR