Sodium propionate ≥99% Kosher

Warning

Synonyms: Propionic acid sodium salt, Sodiumpropionate

W518905-10KG-K W518905-25KG-K W518905-1KG-K
220235-997EA 858.46 USD
220235-997 220235-999 220235-998
Sodium propionate ≥99% Kosher
Sodium propionate
Formula: C₃H₅NaO₂
MW: 96.06 g/mol
Melting Pt: 288.76 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002759
CAS Number: 137-40-6
EINECS: 205-290-4
Merck Index: 12,08816

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR