1,2,3,4-Tetrahydroquinoline

W516007-5KG W516007-1KG W516007-10KG
220235-962EA 80 USD
220235-962 220235-961 220235-960
1,2,3,4-Tetrahydroquinoline
1,2,3,4-Tetrahydroquinoline
Formula: C₉H₁₁N
MW: 133.19 g/mol
MDL Number: MFCD00006693
CAS Number: 635-46-1
EINECS: 211-237-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR