7-Methoxycoumarin

Synonyms: 7-Methoxy-2-benzopyrone, 7-Methoxy-2H-chromen-2-one

W515809-25G W515809-1KG
220235-958EA 201.54 USD
220235-958 220235-957
7-Methoxycoumarin
7-Methoxycoumarin
Formula: C₁₀H₈O₃
MW: 176.17 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 71 °C
Density: 1.36 g/cm³ (20 °C)
MDL Number: MFCD00006876
CAS Number: 531-59-9
EINECS: 208-513-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR