Isopropyl palmitate ≥90%

Synonyms: Hexadecylic acid isopropyl ester, Palmitic acid isopropyl ester, Hexadecanoic acid isopropyl ester, Isopropyl Palmitate, Isopropyl hexadecanoate

W515604-1KG W515604-9KG
220235-948EA 176.92 USD
220235-948 220235-950
Isopropyl palmitate ≥90%
Isopropyl palmitate
Formula: C₁₉H₃₈O₂
MW: 298.51 g/mol
Boiling Pt: 160 °C (2 mmHg)
Melting Pt: 13.5 °C
Density: 0.855 g/cm³ (20 °C)
MDL Number: MFCD00008993
CAS Number: 142-91-6
EINECS: 205-571-1

Order Now

Specification Test Results

Assay 90

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR