2,5-Dimethylthiophene Kosher

W515507-100G-K W515507-25G-K
220235-944EA 80 USD
220235-944 220235-946
2,5-Dimethylthiophene Kosher
2,5-Dimethylthiophene
Formula: C₆H₈S
MW: 112.2 g/mol
Boiling Pt: 134 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.984 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005452
CAS Number: 638-02-8
EINECS: 211-313-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR