4-Methylquinoline ≥99%

Warning

W513903-1KG W513903-100G W513903-5KG
220235-348EA 1053.85 USD
220235-348 220235-347 220235-349
4-Methylquinoline ≥99%
4-Methylquinoline
Formula: C₁₀H₉N
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.0995 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006784
CAS Number: 491-35-0
EINECS: 207-734-2
Merck Index: 13,05460

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR