m-Anisaldehyde ≥97%

Synonyms: 3-Methoxybenzaldehyde

W513105-100G W513105-5KG W513105-1KG
220234-069EA 80 USD
220234-069 220234-926 220234-070
m-Anisaldehyde ≥97%
m-Anisaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 228 - 229 °C (1013 hPa)
Density: 1.1149 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003361
CAS Number: 591-31-1
EINECS: 209-712-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR