3-Pentanone ≥99%

Danger

Synonyms: Diethyl ketone

W510106-1KG W510106-8KG W510106-4KG
220235-817EA 100.92 USD
220235-817 220235-819 220235-818
3-Pentanone ≥99%
3-Pentanone
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 101 °C (1013 hPa)
Melting Pt: -40 °C
Density: 0.8148 g/cm³ (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00009320
CAS Number: 96-22-0
EINECS: 202-490-3
UN: 1156
ADR: 3,II
Merck Index: 13,03148

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR