Methyl palmitate ≥97%

Synonyms: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester

W509531-4KG W509531-1KG W509531-8KG
220235-802EA 516.92 USD
220235-802 220235-801 220235-803
Methyl palmitate ≥97%
Methyl palmitate
Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 417 °C (1013 hPa)
Melting Pt: 29.9 °C
Density: 0.8804 g/cm³ (20 °C)
MDL Number: MFCD00008994
CAS Number: 112-39-0
EINECS: 203-966-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR