Diethylene glycol monoethyl ether ≥99%

Synonyms: 2-(2-Ethoxyethoxy)ethanol, Di(ethylene glycol) ethyl ether, Diethylene Glycol Monoethyl Ether

W509043-1KG W509043-10KG W509043-25KG
220235-790EA 118 USD
220235-790 220235-789 220235-791
Diethylene glycol monoethyl ether ≥99%
Diethylene glycol monoethyl ether
Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: –80 °C
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00002872
CAS Number: 111-90-0
EINECS: 203-919-7
REACH: 01-2119475105-42
Merck Index: 13,01809

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR