3-Methyl-3-pentanol ≥99% Kosher

Synonyms: Diethylmethylcarbinol

W508209-100G-K W508209-4KG-K W508209-1KG-K
220235-774EA 80 USD
220235-774 220235-776 220235-775
3-Methyl-3-pentanol ≥99% Kosher
3-Methyl-3-pentanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 123 °C (1013 hPa)
Melting Pt: -53 °C
Density: 0.8452 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004483
CAS Number: 77-74-7
EINECS: 201-053-4
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR