Xylitol

W507930-1KG W507930-5KG
220235-767EA 216.92 USD
220235-767 220235-768
Xylitol
Xylitol
Formula: C₅H₁₂O₅
Boiling Pt: 495 °C (1013 hPa)
Melting Pt: 94 °C
Density: 1.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064292
CAS Number: 87-99-0
EINECS: 201-788-0
Merck Index: 13,10140

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR