L(-)-Fenchone Kosher

Synonyms: (1R)-(-)-Fenchone

W507709-1KG-K W507709-5KG-K W507709-100G-K
220235-759EA 350.77 USD
220235-759 220235-760 220235-758
L(-)-Fenchone Kosher
L(-)-Fenchone
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: 8.5 °C
Density: 0.962 g/cm³ (0 °C)
MDL Number: MFCD00151104
CAS Number: 7787-20-4
EINECS: 232-107-5
UN: 2717
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR