Methyl stearate ≥96%

Synonyms: Stearic acid methyl ester, Methyl octadecanoate

W504807-1KG W504807-5KG W504807-10KG
220235-715EA 136.77 USD
220235-715 220235-716 220235-714
Methyl stearate ≥96%
Methyl stearate
Formula: CH₃(CH₂)₁₆CO₂CH₃
MW: 298.51 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 1.007 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009005
CAS Number: 112-61-8
EINECS: 203-990-4
Merck Index: 12,08959

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR