2-Methyl-3-buten-2-ol ≥98%

Synonyms: 3-Hydroxy-3-methyl-1-butene

W503908-8KG-K W503908-4KG-K W503908-1KG-K
220235-698EA 80 USD
220235-698 220235-697 220235-696
2-Methyl-3-buten-2-ol ≥98%
2-Methyl-3-buten-2-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 99 °C (1013 hPa)
Melting Pt: –15 °C
Density: 0.824 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004470
CAS Number: 115-18-4
EINECS: 204-068-4
UN: 1987
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR