2,4-Dimethylthiazole

W503401-100G W503401-1KG W503401-25G
220235-686EA 96 USD
220235-686 220235-687 220235-688
2,4-Dimethylthiazole
2,4-Dimethylthiazole
Formula: C₅H₇NS
MW: 113.18 g/mol
Boiling Pt: 144 °C (1013 hPa)
Density: 1.0601 g/cm³ (15 °C)
MDL Number: MFCD00014509
CAS Number: 541-58-2
EINECS: 208-786-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR