Furfuryl butyrate Kosher

W501700-100G-K W501700-5KG-K W501700-1KG-K
220235-665EA 161.54 USD
220235-665 220235-667 220235-666
Furfuryl butyrate Kosher
Furfuryl butyrate
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
CAS Number: 623-21-2
EINECS: 210-779-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR