cis-3-Nonen-1-ol ≥95%

Synonyms: (Z)-3-Nonen-1-ol, cis-3-Nonen-1-ol

W441200-100G W441200-1KG
220234-545EA 76.92 USD
220234-545 220234-546
cis-3-Nonen-1-ol ≥95%
cis-3-Nonen-1-ol
Formula: C₉H₁₈O
MW: 142.24 g/mol
MDL Number: MFCD00010314
CAS Number: 10340-23-5
EINECS: 233-735-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR