Thioacetic acid ≥96%

W421001-1KG W421001-100G
220235-565EA 307.69 USD
220235-565 220235-564
Thioacetic acid ≥96%
Thioacetic acid
Formula: C₂H₄OS
MW: 76.12 g/mol
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: -17 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 18 °C
MDL Number: MFCD00004853
CAS Number: 507-09-5
EINECS: 208-063-8
UN: 2436
ADR: 3,II
Merck Index: 12,09454

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR