(±)-1-Hepten-3-ol ≥97%

Synonyms: 1-Hepten-3-ol, (±)-Hept-1-en-3-ol

W412901-25G W412901-100G W412901-1KG
220235-549EA 261.54 USD
220235-549 220235-547 220235-548
(±)-1-Hepten-3-ol ≥97%
(±)-1-Hepten-3-ol
Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 154 °C (1013 hPa)
Density: 0.8396 g/cm³ (20 °C)
MDL Number: MFCD00021940
CAS Number: 4938-52-7
EINECS: 225-579-9
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR