Diethyl disulfide

Danger

Synonyms: Ethyldisulphide

W409301-100G W409301-1KG W409301-5KG
220235-537EA 184.62 USD
220235-537 220235-538 220235-539
Diethyl disulfide
Diethyl disulphide
Formula: C₄H₁₀S₂
MW: 122.26 g/mol
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: –101.5 °C
Density: 0.984 g/cm³ (20 °C)
MDL Number: MFCD00009266
CAS Number: 110-81-6
EINECS: 203-805-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR