Pyrazine Kosher

W401501-5KG-K W401501-100G-K W401501-1KG-K
220235-512EA 684.62 USD
220235-512 220235-510 220235-511
Pyrazine Kosher
Pyrazine
Formula: C₄H₄N₂
MW: 80.09 g/mol
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: 52 °C
Density: 1.0311 g/cm³ (60.9 °C)
MDL Number: MFCD00006122
CAS Number: 290-37-9
EINECS: 206-027-6
UN: 1325
ADR: 4.1,III
Merck Index: 13,08047

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR