Methyl methacrylate

Danger

Synonyms: Methacrylic acid methyl ester

W400201-10KG W400201-1KG W400201-5KG
220235-491EA 790.77 USD
220235-491 220235-492 220235-493
Methyl methacrylate
Methyl methacrylate
Formula: CH₂=C(CH₃)COOCH₃
MW: 100.12 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: -48 °C
Density: 0.93 g/cm³ (20 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008587
CAS Number: 80-62-6
EINECS: 201-297-1
UN: 1247
ADR: 3,II
REACH: 01-2119452498-28
Merck Index: 12,06005

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR