4-Hydroxybenzyl alcohol ≥98%

Synonyms: (4-Hydroxyphenyl)methanol, p-Hydroxybenzyl Alcohol, 4-(Hydroxymethyl)phenol

W398705-1KG W398705-5KG W398705-100G
220235-471EA 944.62 USD
220235-471 220235-472 220235-470
4-Hydroxybenzyl alcohol ≥98%
4-Hydroxybenzyl alcohol
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: 115 - 118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004658
CAS Number: 623-05-2
EINECS: 210-768-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR