Phenyl salicylate ≥99%

Warning

Synonyms: Phenyl 2-hydroxybenzoate, Phenyl Salicylate

W396001-1KG W396001-5KG
220235-320EA 210.77 USD
220235-320 220235-321
Phenyl salicylate ≥99%
Phenyl salicylate
Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 41…43 °C
Density: 1.307 g/cm³ (20 °C)
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002213
CAS Number: 118-55-8
EINECS: 204-259-2
UN: 3077
ADR: 9,III
Merck Index: 12,07464

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR