Biphenyl-2-ol ≥99%

Warning

Synonyms: 2-Phenylphenol, 2-Hydroxydiphenyl, 2-Hydroxybiphenyl

W395900-5KG W395900-1KG W395900-10KG
220234-417EA 80 USD
220234-417 220234-416 220234-415
Biphenyl-2-ol ≥99%
Biphenyl-2-ol
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 280 - 286 °C (1013 hPa)
Melting Pt: 56 - 58 °C
Density: 1.213 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00002208
CAS Number: 90-43-7
EINECS: 201-993-5
UN: 3077
ADR: 9,III
Merck Index: 12,07458

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR