trans,trans-2,4-Hexadien-1-ol Kosher

Synonyms: (2E,4E)-Hexa-2,4-dien-1-ol, Trans,trans-2,4-hexadien-1-ol, (2E,4E)-1-Hydroxyhexa-2,4-diene, Sorbic alcohol, trans,trans-Hexa-2,4-dien-1-ol

W392200-1KG-K W392200-100G-K W392200-25G-K
220235-309EA 633.85 USD
220235-309 220235-308 220235-310
trans,trans-2,4-Hexadien-1-ol Kosher
trans,trans-2,4-Hexadien-1-ol
Formula: C₆H₁₀O
MW: 98.14 g/mol
MDL Number: MFCD00002925
CAS Number: 17102-64-6
EINECS: 241-173-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR