4-tert-Butylphenol ≥99%

W391808-1KG W391808-9KG W391808-20KG
220235-298EA 633.85 USD
220235-298 220235-300 220235-299
4-tert-Butylphenol ≥99%
4-tert-Butylphenol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 95…98 °C
Density: 1.017 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002367
CAS Number: 98-54-4
EINECS: 202-679-0
UN: 2430
ADR: 8,III
Merck Index: 13,01584

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR