Cyclohexanone ≥99.8%

Danger

W390909-1KG W390909-10KG W390909-25KG
220235-290EA 100.92 USD
220235-290 220235-289 220235-291
Cyclohexanone ≥99.8%
Cyclohexanone
Formula: C₆H₁₀(=O)
Boiling Pt: 155.6 °C (1013 hPa)
Melting Pt: -31 °C
Density: 0.9478 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3,III
REACH: 01-2119453616-35
Merck Index: 13,02755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR