3-Methyl-1-butanethiol

Synonyms: 1-Mercapto-3-methylbutane, 3-Methylbutyl mercaptan, Isopentyl mercaptan

W385808-4KG W385808-100G W385808-1KG
220235-243EA 1135.38 USD
220235-243 220235-241 220235-242
3-Methyl-1-butanethiol
3-Methyl-1-butanethiol
Formula: C₅H₁₂S
MW: 104.22 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –134 °C
Density: 0.8548 g/cm³ (0 °C)
MDL Number: MFCD00004899
CAS Number: 541-31-1
EINECS: 208-774-3
UN: 1111
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR