Diisopropyl disulfide ≥96%

Synonyms: Isopropyl disulphide, 1,2-Diisopropyldisulphane

W382701-100G W382701-5KG W382701-1KG
220235-213EA 204.62 USD
220235-213 220235-215 220235-214
Diisopropyl disulfide ≥96%
Diisopropyl disulphide
Formula: C₆H₁₄S₂
MW: 150.31 g/mol
Boiling Pt: 175 °C (1013 hPa)
Melting Pt: –69 °C
Density: 0.943 g/cm³ (20 °C)
MDL Number: MFCD00008894
CAS Number: 4253-89-8
EINECS: 224-225-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR