L(+)-Arginine ≥98.5%

Warning

Synonyms: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH

W381918-1KG W381918-10KG W381918-5KG
220235-193EA 148 USD
220235-193 220235-192 220235-194
L(+)-Arginine ≥98.5%
L(+)-Arginine
Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 238 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR