DL-Alanine ≥99% Kosher

Synonyms: (±)-Alanine, DL-α-Alanine, H-DL-Ala-OH, α-Alanine

W381810-10KG-K W381810-5KG-K W381810-1KG-K
220235-185EA 941.54 USD
220235-185 220235-187 220235-186
DL-Alanine ≥99% Kosher
DL-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064408
CAS Number: 302-72-7
EINECS: 206-126-4
Merck Index: 13,00203

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR